Learn More
Fluotics Analytik Jena Cybi Well compatible P60, sterile
Catalog No. 5000435126
Encompass_Preferred
Analytik Jena Cybi Well compatible P60, sterile, Working Volume: 60uL, Tip Color: Clear, Tip Tray, 384/60 uL, Compare To: OL3800-25-715-P, 24 racks/case with 384 tips per rack
Catalog No. 50-004-35126 Supplier Fluotics Supplier No. CYBW60.ST
Analytik Jena Cybi Well compatible P60, sterile, Working Volume: 60uL, Tip Color: Clear, Tip Tray, 384/60 uL, Compare To: OL3800-25-715-P, 24 racks/case with 384 tips per rack