Learn More
Fluotics Molecular Devices FLIPR Compatible P30, non-sterile
Catalog No. 5000435166
Encompass_Preferred
Molecular Devices FLIPR Compatible P30, non-sterile, Working Volume: 30uL, Tip Color: Clear, Tip Box, FLIPR 384 Head, Compare To: 9000-0763, 50 racks/case with 384 tips per rack
Catalog No. 50-004-35166 Supplier Fluotics Supplier No. FLP30.NS
Molecular Devices FLIPR Compatible P30, non-sterile, Working Volume: 30uL, Tip Color: Clear, Tip Box, FLIPR 384 Head, Compare To: 9000-0763, 50 racks/case with 384 tips per rack