Learn More
Fluotics Analytik Jena Cybio FeliX compatible P10, sterile
Supplier:  Fluotics CYBF10TT.ST
Encompass_Preferred
Analytik Jena Cybio FeliX compatible P10, sterile, Working Volume: 10uL, Tip Color: Clear, Tip Tray, R 384, 24 racks/case with 384 tips/rack
Catalog No. 50-004-35163