Learn More
Fluotics Analytik Jena Cybio FeliX compatible P10, non-sterile
Supplier:  Fluotics CYBF10TT.NS
Encompass_Preferred
Analytik Jena Cybio FeliX compatible P10, non-sterile, Working Volume: 10uL, Tip Color: Clear, Tip Tray, R 384, 24 racks/case with 384 tips/rack
Catalog No. 50-004-35177