Learn More
Fluotics Analytik Jena Cybio FeliX compatible P1000, sterile
Catalog No. 5000435160
Encompass
Analytik Jena Cybio FeliX compatible P1000, sterile, Working Volume: 1000uL, Tip Color: Clear, Tip Tray, Compare To: OL3810-25-874-P, 24 racks/case with 96 tips/rack
Catalog No. 50-004-35160 Supplier Fluotics Supplier No. CYBF1000TT.ST
Analytik Jena Cybio FeliX compatible P1000, sterile, Working Volume: 1000uL, Tip Color: Clear, Tip Tray, Compare To: OL3810-25-874-P, 24 racks/case with 96 tips/rack