Learn More
Fluotics Analytik Jena Cybio FeliX compatible P1000, filter/sterile
Catalog No. 5000435270
Encompass
Analytik Jena Cybio FeliX compatible P1000, filter/sterile, Working Volume: 950uL, Tip Color: Clear, Tip Tray, Compare To: OL3810-25-879-F, 24 racks/case with 96 tips/rack
Catalog No. 50-004-35270 Supplier Fluotics Supplier No. CYBF1000TT.F
Analytik Jena Cybio FeliX compatible P1000, filter/sterile, Working Volume: 950uL, Tip Color: Clear, Tip Tray, Compare To: OL3810-25-879-F, 24 racks/case with 96 tips/rack