Learn More
Fluotics Analytik Jena Cybio FeliX compatible P250, filter/sterile
Catalog No. 5000435269
Encompass
Analytik Jena Cybio FeliX compatible P250, filter/sterile, Working Volume: 200uL, Tip Color: Clear, Tip Tray, R 96/250 uL, Compare To: OL3810-25-669, 24 robotrays/case with 96 tips per tray
Catalog No. 50-004-35269 Supplier Fluotics Supplier No. CYBF250TT.F
Add to Cart
Add to Cart
Analytik Jena Cybio FeliX compatible P250, filter/sterile, Working Volume: 200uL, Tip Color: Clear, Tip Tray, R 96/250 uL, Compare To: OL3810-25-669, 24 robotrays/case with 96 tips per tray