Learn More
Fluotics Analytik Jena Cybio FeliX compatible P40, filter/sterile, Tips in Quadrant only
Catalog No. 5000435148
Encompass
Analytik Jena Cybio FeliX compatible P40, filter/sterile, Tips in Quadrant only, Working Volume: 35uL, Tip Color: Clear, Tip Tray, R 384 w/ adapter, 24 robotrays/case with 96 tips per tray
Catalog No. 50-004-35148 Supplier Fluotics Supplier No. CYBF40Q.F
Analytik Jena Cybio FeliX compatible P40, filter/sterile, Tips in Quadrant only, Working Volume: 35uL, Tip Color: Clear, Tip Tray, R 384 w/ adapter, 24 robotrays/case with 96 tips per tray