Learn More
Fluotics Analytik Jena Cybio FeliX compatible P40, sterile, Tips in Quadrant only
Catalog No. 5000435206
Encompass
Analytik Jena Cybio FeliX compatible P40, sterile, Tips in Quadrant only, Working Volume: 40uL, Tip Color: Clear, Tip Tray, R 384 w/ adapter, Compare To: OL3810-25-434, 24 robotrays/case with 96 tips per tray
Catalog No. 50-004-35206 Supplier Fluotics Supplier No. CYBF40Q.ST
Analytik Jena Cybio FeliX compatible P40, sterile, Tips in Quadrant only, Working Volume: 40uL, Tip Color: Clear, Tip Tray, R 384 w/ adapter, Compare To: OL3810-25-434, 24 robotrays/case with 96 tips per tray