Learn More
Fluotics Analytik Jena Cybio FeliX compatible P50, filter/sterile
Catalog No. 5000435267
Encompass
Analytik Jena Cybio FeliX compatible P50, filter/sterile, Working Volume: 50uL, Tip Color: Clear, Tip Box, SELECT/CHOICE 96, Compare To: 30-102-860, OL3811-25-935-F, 50 racks/case with 96 tips per rack
Catalog No. 50-004-35267 Supplier Fluotics Supplier No. CYBF50.F
Add to Cart
Add to Cart
Analytik Jena Cybio FeliX compatible P50, filter/sterile, Working Volume: 50uL, Tip Color: Clear, Tip Box, SELECT/CHOICE 96, Compare To: 30-102-860, OL3811-25-935-F, 50 racks/case with 96 tips per rack