Learn More
Fluotics Analytik Jena Cybio FeliX compatible P40, filter/sterile
Catalog No. 5000435241
Encompass_Preferred
Analytik Jena Cybio FeliX compatible P40, filter/sterile, Working Volume: 35uL, Tip Color: Clear, Tip Tray, R 384, 24 robotrays/case with 384 tips per tray
Catalog No. 50-004-35241 Supplier Fluotics Supplier No. CYBF40TT.F
Analytik Jena Cybio FeliX compatible P40, filter/sterile, Working Volume: 35uL, Tip Color: Clear, Tip Tray, R 384, 24 robotrays/case with 384 tips per tray