Learn More
Fluotics Analytik Jena Cybio FeliX compatible P50, sterile
Catalog No. 5000435242
Encompass
Analytik Jena Cybio FeliX compatible P50, sterile, Working Volume: 50uL, Tip Color: Clear, Tip Box, SELECT/CHOICE 96, Compare To: 30-102-858, OL3811-25-635-S, 50 racks/case with 96 tips per rack
Catalog No. 50-004-35242 Supplier Fluotics Supplier No. CYBF50.ST
Analytik Jena Cybio FeliX compatible P50, sterile, Working Volume: 50uL, Tip Color: Clear, Tip Box, SELECT/CHOICE 96, Compare To: 30-102-858, OL3811-25-635-S, 50 racks/case with 96 tips per rack