Learn More
Fluotics Analytik Jena Cybio FeliX compatible P60, sterile
Catalog No. 5000435149
Encompass_Preferred
Analytik Jena Cybio FeliX compatible P60, sterile, Working Volume: 60uL, Tip Color: Clear, Tip Tray, R 384, Compare To: OL3810-25-244, 24 robotray/case with 384 tips per tray
Catalog No. 50-004-35149 Supplier Fluotics Supplier No. CYBF60TT.ST
Analytik Jena Cybio FeliX compatible P60, sterile, Working Volume: 60uL, Tip Color: Clear, Tip Tray, R 384, Compare To: OL3810-25-244, 24 robotray/case with 384 tips per tray