Learn More
Fluotics Analytik Jena Cybio FeliX compatible P60, sterile, Tips in Quadrant only
Catalog No. 5000435226
Encompass
Analytik Jena Cybio FeliX compatible P60, sterile, Tips in Quadrant only, Working Volume: 60uL, Tip Color: Clear, Tip Tray, R 384 w/ adapter, Compare To: OL3810-25-444, 24 robotrays/case with 96 tips per tray
Catalog No. 50-004-35226 Supplier Fluotics Supplier No. CYBF60Q.ST
Analytik Jena Cybio FeliX compatible P60, sterile, Tips in Quadrant only, Working Volume: 60uL, Tip Color: Clear, Tip Tray, R 384 w/ adapter, Compare To: OL3810-25-444, 24 robotrays/case with 96 tips per tray