Learn More
Fluotics Analytik Jena Cybio FeliX compatible P60, filter/sterile, Tips in Quadrant only
Catalog No. 5000435271
Encompass
Analytik Jena Cybio FeliX compatible P60, filter/sterile, Tips in Quadrant only, Working Volume: 45uL, Tip Color: Clear, Tip Tray, R 384 w/ adapter, Compare To: OL3810-25-449, 24 robotrays/case with 96 tips per tray
Catalog No. 50-004-35271 Supplier Fluotics Supplier No. CYBF60Q.F
Analytik Jena Cybio FeliX compatible P60, filter/sterile, Tips in Quadrant only, Working Volume: 45uL, Tip Color: Clear, Tip Tray, R 384 w/ adapter, Compare To: OL3810-25-449, 24 robotrays/case with 96 tips per tray