Learn More
Fluotics Analytik Jena Cybio FeliX compatible P60, filter/sterile
Catalog No. 5000435243
Encompass_Preferred
Analytik Jena Cybio FeliX compatible P60, filter/sterile, Working Volume: 45uL, Tip Color: Clear, Tip Tray, R 384, Compare To: OL3810-25-249, 24 robotrays/case with 384 tips per tray
Catalog No. 50-004-35243 Supplier Fluotics Supplier No. CYBF60TT.F
Analytik Jena Cybio FeliX compatible P60, filter/sterile, Working Volume: 45uL, Tip Color: Clear, Tip Tray, R 384, Compare To: OL3810-25-249, 24 robotrays/case with 384 tips per tray