Learn More
Litron Laboratories ANALYZE 60 RAT BLOOD SAMPLES
Catalog No. NC2595051
Encompass
  • Engineered for flow cytometric analysis of fixed rat blood samples.
  • Allows analysis of 60 rat blood samples simultaneously.
  • Compatible with 488 nm excitation flow cytometers.
  • Includes quality-assured reagents for in-house analyses.
  • Offers evaluation of micronuclei, young red blood cells (CD71), and platelets (CD61).
  • Streamlines workflow with solutions for washing, RNA removal, DNA staining, and calibration.
  • Requires previously fixed and frozen (-80 °C) rat blood samples.
  • Stores components at -80 °C, -20 °C, 4 °C, and ambient temperatures.
  • Requires external equipment like centrifuge and flow cytometer.
  • Compatible with BD CellQuest™ Pro and FACSDiva™ software.
Catalog No. NC2595051 Supplier Litron Laboratories Supplier No. PLUSR
  • Engineered for flow cytometric analysis of fixed rat blood samples.
  • Allows analysis of 60 rat blood samples simultaneously.
  • Compatible with 488 nm excitation flow cytometers.
  • Includes quality-assured reagents for in-house analyses.
  • Offers evaluation of micronuclei, young red blood cells (CD71), and platelets (CD61).
  • Streamlines workflow with solutions for washing, RNA removal, DNA staining, and calibration.
  • Requires previously fixed and frozen (-80 °C) rat blood samples.
  • Stores components at -80 °C, -20 °C, 4 °C, and ambient temperatures.
  • Requires external equipment like centrifuge and flow cytometer.
  • Compatible with BD CellQuest™ Pro and FACSDiva™ software.