Learn More
Alkali Scientific Analyzer Cuvette for Hitachi 902 Analyzer 6/Pk, 24/Case
SDP
Catalog No. 5000661937
Encompass_Preferred
This analyzer cuvette is specifically designed for use with the Hitachi 902 analyzer and manufactured to meet strict dimensional and optical requirements for clinical chemistry applications. The cuvette ensures proper alignment within the analyzer, consistent light transmission, and reliable photometric readings during routine and high throughput testing. Produced from high quality optically clear plastic, each cuvette provides uniform wall thickness and a consistent optical path length to support accurate and reproducible results. The smooth surface finish helps reduce carryover and minimizes interference during sample analysis, contributing to dependable day to day instrument performance. Suitable for clinical laboratories, hospitals, and diagnostic facilities, these cuvettes serve as a reliable consumable for routine chemistry testing workflows.
Catalog No. 50-006-61937 Supplier Alkali Scientific Supplier No. HAC902
This analyzer cuvette is specifically designed for use with the Hitachi 902 analyzer and manufactured to meet strict dimensional and optical requirements for clinical chemistry applications. The cuvette ensures proper alignment within the analyzer, consistent light transmission, and reliable photometric readings during routine and high throughput testing. Produced from high quality optically clear plastic, each cuvette provides uniform wall thickness and a consistent optical path length to support accurate and reproducible results. The smooth surface finish helps reduce carryover and minimizes interference during sample analysis, contributing to dependable day to day instrument performance. Suitable for clinical laboratories, hospitals, and diagnostic facilities, these cuvettes serve as a reliable consumable for routine chemistry testing workflows.