Learn More
Apexbio Technology LLC Anamorelin 249921-19-5 100mg
SDP
Catalog No. 5000095197
Encompass_Preferred
Anamorelin (CAS 249921-19-5) is a synthetic small-molecule agonist of the growth hormone secretagogue receptor (GHSR) By binding to and activating GHSR centrally Anamorelin replicates the orexigenic and anabolic effects of endogenous ghrelin promoting appetite stimulation and growth hormone release This mechanism may also contribute to modulation of inflammatory pathways as GHSR activation is associated with reduced production of pro-inflammatory cytokines such as TNF-alpha and interleukin-6 Anamorelin is primarily investigated for its potential in studies of cancer cachexia appetite regulation and metabolic research
Catalog No. 50-000-95197 Supplier Apexbio Technology LLC Supplier No. A3179100
Anamorelin (CAS 249921-19-5) is a synthetic small-molecule agonist of the growth hormone secretagogue receptor (GHSR) By binding to and activating GHSR centrally Anamorelin replicates the orexigenic and anabolic effects of endogenous ghrelin promoting appetite stimulation and growth hormone release This mechanism may also contribute to modulation of inflammatory pathways as GHSR activation is associated with reduced production of pro-inflammatory cytokines such as TNF-alpha and interleukin-6 Anamorelin is primarily investigated for its potential in studies of cancer cachexia appetite regulation and metabolic research