Learn More
Medchemexpress LLC Anamorelin | 249921-19-5 | 546.70 | 5 MG
SDP
Catalog No. 5000359637
Encompass_Preferred
Anamorelin (RC-1291) is an orally active Ghrelin receptor agonist. It promotes appetite, increases body weight, and stimulates the secretion of growth hormone and insulin-like growth factor-1. Anamorelin can be used in the research of anorexia and cancer cachexia.
  • Orally active Ghrelin receptor agonist
  • Promotes appetite
  • Increases body weight
  • Stimulates growth hormone secretion
  • Stimulates insulin-like growth factor-1 secretion
  • Used in anorexia research
  • Used in cancer cachexia research
Catalog No. 50-003-59637 Supplier Medchemexpress LLC Supplier No. HY147345MG
Anamorelin (RC-1291) is an orally active Ghrelin receptor agonist. It promotes appetite, increases body weight, and stimulates the secretion of growth hormone and insulin-like growth factor-1. Anamorelin can be used in the research of anorexia and cancer cachexia.
  • Orally active Ghrelin receptor agonist
  • Promotes appetite
  • Increases body weight
  • Stimulates growth hormone secretion
  • Stimulates insulin-like growth factor-1 secretion
  • Used in anorexia research
  • Used in cancer cachexia research