Learn More
Medchemexpress LLC Anamorelin | 249921-19-5 | 99.9% | 546.70 | 100 MG
SDP
Catalog No. 5000360384
Encompass_Preferred
Anamorelin is an orally active Ghrelin receptor agonist with an EC50 of 0.74 nM. It promotes appetite, increases body weight, and stimulates the secretion of growth hormone and insulin-like growth factor-1. It is used in the research of anorexia and cancer cachexia.
  • Orally active Ghrelin receptor agonist
  • Promotes appetite
  • Increases body weight
  • Stimulates growth hormone secretion
  • Stimulates insulin-like growth factor-1 secretion
  • Used in anorexia research
  • Used in cancer cachexia research
Catalog No. 50-003-60384 Supplier Medchemexpress LLC Supplier No. HY14734100MG
Anamorelin is an orally active Ghrelin receptor agonist with an EC50 of 0.74 nM. It promotes appetite, increases body weight, and stimulates the secretion of growth hormone and insulin-like growth factor-1. It is used in the research of anorexia and cancer cachexia.
  • Orally active Ghrelin receptor agonist
  • Promotes appetite
  • Increases body weight
  • Stimulates growth hormone secretion
  • Stimulates insulin-like growth factor-1 secretion
  • Used in anorexia research
  • Used in cancer cachexia research