Learn More
Selleck Chemical LLC Anamorelin S4980-10mM/1mL
SDP
Catalog No. 503110722
Encompass_Preferred
Anamorelin (ONO-7643 RC-1291 ST-1291) is an orally active high-affinity selective agonist of the ghrelin receptor with an EC50 value of 0 74 nM in the HEK293/GRLN FLIPR assay
Catalog No. 50-311-0722 Supplier Selleck Chemical LLC Supplier No. S498010MM/1ML
Anamorelin (ONO-7643 RC-1291 ST-1291) is an orally active high-affinity selective agonist of the ghrelin receptor with an EC50 value of 0 74 nM in the HEK293/GRLN FLIPR assay