Learn More
Medchemexpress LLC Anamorelin (hydrochloride) | 861998-00-7 | 99.6% | 583.16 | 1 ML
SDP
Catalog No. 5000361437
Encompass_Preferred
Anamorelin hydrochloride is an orally active Ghrelin receptor agonist with an EC50 of 0.74 nM. It promotes appetite, increases body weight, and stimulates the secretion of growth hormone and insulin-like growth factor-1. It is suitable for research into anorexia and cancer cachexia.
  • Orally active Ghrelin receptor agonist
  • Promotes appetite
  • Increases body weight
  • Stimulates growth hormone and insulin-like growth factor-1 secretion
  • Used in research of anorexia
  • Used in research of cancer cachexia
Catalog No. 50-003-61437 Supplier Medchemexpress LLC Supplier No. HY14734A10MM/1ML
Add to Cart
Add to Cart
Anamorelin hydrochloride is an orally active Ghrelin receptor agonist with an EC50 of 0.74 nM. It promotes appetite, increases body weight, and stimulates the secretion of growth hormone and insulin-like growth factor-1. It is suitable for research into anorexia and cancer cachexia.
  • Orally active Ghrelin receptor agonist
  • Promotes appetite
  • Increases body weight
  • Stimulates growth hormone and insulin-like growth factor-1 secretion
  • Used in research of anorexia
  • Used in research of cancer cachexia