Learn More
Medchemexpress LLC Anamorelin hydrochloride | 861998-00-7 | 99.6% | 583.16 | 50 MG
SDP
Catalog No. 5000360373
Encompass_Preferred
Anamorelin hydrochloride is an orally active Ghrelin receptor agonist with an EC50 of 0.74 nM. This compound can promote appetite, increase body weight, and stimulate the secretion of growth hormone and insulin-like growth factor-1. It is suitable for research into anorexia and cancer cachexia.
Solid: 4°C, sealed storage, away from moisture.
In solvent: -80°C for 6 months; -20°C for 1 month (sealed storage, away from moisture).
  • Orally active Ghrelin receptor agonist.
  • EC50 of 0.74 nM for the Ghrelin receptor.
  • Promotes appetite and increases body weight.
  • Stimulates growth hormone and insulin-like growth factor-1 secretion.
Catalog No. 50-003-60373 Supplier Medchemexpress LLC Supplier No. HY14734A50MG
Anamorelin hydrochloride is an orally active Ghrelin receptor agonist with an EC50 of 0.74 nM. This compound can promote appetite, increase body weight, and stimulate the secretion of growth hormone and insulin-like growth factor-1. It is suitable for research into anorexia and cancer cachexia.
Solid: 4°C, sealed storage, away from moisture.
In solvent: -80°C for 6 months; -20°C for 1 month (sealed storage, away from moisture).
  • Orally active Ghrelin receptor agonist.
  • EC50 of 0.74 nM for the Ghrelin receptor.
  • Promotes appetite and increases body weight.
  • Stimulates growth hormone and insulin-like growth factor-1 secretion.