Learn More
Medchemexpress LLC Anamorelin hydrochloride | 861998-00-7 | MFCD23098788 | 99.59% | 583.16 | 100 MG
SDP
Catalog No. 5000359979
Encompass_Preferred
Anamorelin hydrochloride is an orally active, small-molecule ghrelin receptor agonist. It promotes appetite, increases body weight, and stimulates the secretion of growth hormone and insulin-like growth factor-1. This compound is used in the research of anorexia and cancer cachexia, mimicking ghrelin's effects and potentially decreasing pro-inflammatory cytokines.
  • Orally active ghrelin receptor agonist with an EC50 of 0.74 nM
  • Promotes appetite and increases body weight
  • Stimulates secretion of growth hormone and insulin-like growth factor-1
  • Used in research for anorexia and cancer cachexia
  • Orally bioavailable hydrochloride salt
  • Functions by binding and stimulating the growth hormone secretagogue receptor (GHSR)
  • May decrease pro-inflammatory cytokines such as TNF-alpha and interleukin-6
Catalog No. 50-003-59979 Supplier Medchemexpress LLC Supplier No. HY14734A100MG
Anamorelin hydrochloride is an orally active, small-molecule ghrelin receptor agonist. It promotes appetite, increases body weight, and stimulates the secretion of growth hormone and insulin-like growth factor-1. This compound is used in the research of anorexia and cancer cachexia, mimicking ghrelin's effects and potentially decreasing pro-inflammatory cytokines.
  • Orally active ghrelin receptor agonist with an EC50 of 0.74 nM
  • Promotes appetite and increases body weight
  • Stimulates secretion of growth hormone and insulin-like growth factor-1
  • Used in research for anorexia and cancer cachexia
  • Orally bioavailable hydrochloride salt
  • Functions by binding and stimulating the growth hormone secretagogue receptor (GHSR)
  • May decrease pro-inflammatory cytokines such as TNF-alpha and interleukin-6