Learn More
Medchemexpress LLC Anamorelin (RC-1291) | 249921-19-5 | 99.9% | 546.70 | 200 MG
SDP
Catalog No. 5000362956
Encompass_Preferred
Anamorelin (RC-1291) is an orally active Ghrelin receptor agonist with an EC50 of 0.74 nM. It promotes appetite, increases body weight, and stimulates the secretion of growth hormone and insulin-like growth factor-1. Anamorelin can be used in the research of anorexia and cancer cachexia.
  • Orally active Ghrelin receptor agonist with an EC50 of 0.74 nM
  • Promotes appetite and increases body weight
  • Stimulates secretion of growth hormone and insulin-like growth factor-1
  • Used in research of anorexia and cancer cachexia
  • Stimulates growth hormone release in rat pituitary cells
  • Does not promote tumor growth in lung cancer xenograft models
Catalog No. 50-003-62956 Supplier Medchemexpress LLC Supplier No. HY14734200MG
Anamorelin (RC-1291) is an orally active Ghrelin receptor agonist with an EC50 of 0.74 nM. It promotes appetite, increases body weight, and stimulates the secretion of growth hormone and insulin-like growth factor-1. Anamorelin can be used in the research of anorexia and cancer cachexia.
  • Orally active Ghrelin receptor agonist with an EC50 of 0.74 nM
  • Promotes appetite and increases body weight
  • Stimulates secretion of growth hormone and insulin-like growth factor-1
  • Used in research of anorexia and cancer cachexia
  • Stimulates growth hormone release in rat pituitary cells
  • Does not promote tumor growth in lung cancer xenograft models