Learn More
Aviva Systems Biology Corp ANAPC10 Antibody - N-terminal region (ARP43169_P050) | 100uL
SDP
Catalog No. 5001458454
Encompass_Preferred
This is a rabbit polyclonal antibody against ANAPC10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ANAPC10. Alternate names: APC10, DKFZP564L0562, DOC1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58454 Supplier Aviva Systems Biology Corp Supplier No. ARP43169_P050
This is a rabbit polyclonal antibody against ANAPC10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ANAPC10. Alternate names: APC10, DKFZP564L0562, DOC1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish