Learn More
Aviva Systems Biology Corp ANAPC7 Antibody - C-terminal region (ARP41605_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41605_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ANAPC7. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human ANAPC7. Alternate names: APC7ApplicationIHC, WB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53310