Learn More
Biochain Institute Inc AnaPrep Blood DNA Extraction Kit 200 (48) For extracting genomic DNA from mammalian whole blood, peripheral mononuclear cell, or buffy coat. Sample volume range: up to 400 ml, 1 kit (48 extractions)/pk
SDP
Catalog No. 501871573
Encompass_Preferred
The AnaPrep Blood DNA Extraction Kit is for use with the AnaPrep 12 instruments for extraction of DNA from100‐400 μl mammalian whole blood and buffy coat. For Research Use Only. Not for use in diagnostics procedures.
Catalog No. 50-187-1573 Supplier Biochain Institute Inc Supplier No. Z1322001
The AnaPrep Blood DNA Extraction Kit is for use with the AnaPrep 12 instruments for extraction of DNA from100‐400 μl mammalian whole blood and buffy coat. For Research Use Only. Not for use in diagnostics procedures.