Learn More
Biochain Institute Inc AnaPrep FFPE DNA Extraction Kit (48) For extracting genomic DNA from formalin-fixed paraffin-embedded tissue (FFPE) samples, 1 kit (48 extractions)/pk
SDP
Catalog No. 501871579
Encompass_Preferred
AnaPrep FFPE DNA Extraction Kit is used with the AnaPrep 12 instrument for extraction of genomic DNA from FFPE (Formalin‐Fixed, Paraffin‐Embedded) tissue samples. Providing good quality, high integrity DNA for Molecular diagnosis and research. For Research Use Only. Not for use in diagnostics procedures.
Catalog No. 50-187-1579 Supplier Biochain Institute Inc Supplier No. Z1322009
AnaPrep FFPE DNA Extraction Kit is used with the AnaPrep 12 instrument for extraction of genomic DNA from FFPE (Formalin‐Fixed, Paraffin‐Embedded) tissue samples. Providing good quality, high integrity DNA for Molecular diagnosis and research. For Research Use Only. Not for use in diagnostics procedures.