Learn More
Biochain Institute Inc AnaPrep HPV DNA Extraction Kit for swab samples (48) For extracting HPV DNA from swab samples, 1 kit (48 extractions)/pk
SDP
Catalog No. 501871577
Encompass_Preferred
The AnaPrep HPV DNA Extraction Kit is used with the AnaPrep 12 instrument for DNA extraction of the Human Papillomavirus (HPV) from cervical cell samples which collected by cervical brush or genital swab in liquid‐based Medium* (e.g. Hologic Thinprep PreservCyt , BD SurepathTM, etc.) or other STM (sample transport media) preservation solutions (e.g. QIAGEN DNA PAP Cervical sampler, Roche Cobas PCR Cell Collection Media, Hybribio cell preservation solution, etc.). *The liquid‐base medium are formulated for cellular preservation and used in liquid‐based cytological systems(LBC) for cytological and molecular diagnosis. For Research Use Only. Not for use in diagnostics procedures.
Catalog No. 50-187-1577 Supplier Biochain Institute Inc Supplier No. Z1322007
The AnaPrep HPV DNA Extraction Kit is used with the AnaPrep 12 instrument for DNA extraction of the Human Papillomavirus (HPV) from cervical cell samples which collected by cervical brush or genital swab in liquid‐based Medium* (e.g. Hologic Thinprep PreservCyt , BD SurepathTM, etc.) or other STM (sample transport media) preservation solutions (e.g. QIAGEN DNA PAP Cervical sampler, Roche Cobas PCR Cell Collection Media, Hybribio cell preservation solution, etc.). *The liquid‐base medium are formulated for cellular preservation and used in liquid‐based cytological systems(LBC) for cytological and molecular diagnosis. For Research Use Only. Not for use in diagnostics procedures.