Learn More
Biochain Institute Inc AnaPrep Tissue DNA Extraction Kit (48) For extracting genomic DNA from a variety of animal tissues, swab, and blood stains, 1 kit (48 extractions)/pk
SDP
Catalog No. 501871575
Encompass_Preferred
AnaPrep Tissue DNA Extraction Kit is used with the AnaPrep 12 instruments for extraction of genomic DNA from a varietyof animal tissues, swab samples and blood stains. For extraction from FFPE samples, using “Z1322009 AnaPrep FFPE DNA extraction kit” is recommended. For Research Use Only. Not for use in diagnostics procedures.
Catalog No. 50-187-1575 Supplier Biochain Institute Inc Supplier No. Z1322004
AnaPrep Tissue DNA Extraction Kit is used with the AnaPrep 12 instruments for extraction of genomic DNA from a varietyof animal tissues, swab samples and blood stains. For extraction from FFPE samples, using “Z1322009 AnaPrep FFPE DNA extraction kit” is recommended. For Research Use Only. Not for use in diagnostics procedures.