Learn More
Biochain Institute Inc AnaPrep Viral Nucleic Acid Extraction Kit (48) For extracting viral nucleic acids from plasma, 1 kit (48 extractions)/pk
SDP
Catalog No. 501871574
Encompass_Preferred
The AnaPrep Viral Nucleic Acid Extraction Kit is used with the AnaPrep 12 instrument for extraction of Viral DNA orRNA from human biological specimens such as serum, plasma, and other cell‐free fluids. For Research Use Only. Not for use in diagnostics procedures.
Catalog No. 50-187-1574 Supplier Biochain Institute Inc Supplier No. Z1322003
The AnaPrep Viral Nucleic Acid Extraction Kit is used with the AnaPrep 12 instrument for extraction of Viral DNA orRNA from human biological specimens such as serum, plasma, and other cell‐free fluids. For Research Use Only. Not for use in diagnostics procedures.