Learn More
Biochain Institute Inc AnaPrep Viral/Pathogen Nucleic Acids Extraction Kit A, 1 kit/pk
SDP
Catalog No. 501871581
Encompass_Preferred
The AnaPrep Viral/Pathogen Nucleic Acids Extraction Kit A is used with the AnaPrep 12 instrument forextraction of Viral DNA or RNA from cell-free samples such as serum, plasma, and other cell-free bodyfluids. For Research Use Only. Not for use in diagnostics procedures.
Catalog No. 50-187-1581 Supplier Biochain Institute Inc Supplier No. Z1322011
The AnaPrep Viral/Pathogen Nucleic Acids Extraction Kit A is used with the AnaPrep 12 instrument forextraction of Viral DNA or RNA from cell-free samples such as serum, plasma, and other cell-free bodyfluids. For Research Use Only. Not for use in diagnostics procedures.