Learn More
Biochain Institute Inc AnaPrep Viral/Pathogen Nucleic Acids Extraction Kit B, 1 kit/pk
SDP
Catalog No. 501871582
Encompass_Preferred
The AnaPrep Viral/Pathogen Nucleic Acids Extraction Kit B is used with the AnaPrep 12 instrument forextraction of Viral DNA or RNA from swab samples (cell-rich samples). For Research Use Only. Not for use in diagnostics procedures.
Catalog No. 50-187-1582 Supplier Biochain Institute Inc Supplier No. Z1322012
The AnaPrep Viral/Pathogen Nucleic Acids Extraction Kit B is used with the AnaPrep 12 instrument forextraction of Viral DNA or RNA from swab samples (cell-rich samples). For Research Use Only. Not for use in diagnostics procedures.