Learn More
Medchemexpress LLC HY-14274 10mg Medchemexpress, Anastrozole CAS:120511-73-1 Purity:>98%
SDP
Catalog No. 501969896
Encompass_Preferred
Medchemexpress, HY-14274 10mg Anastrozole CAS:120511-73-1 Anastrozole is a potent, highly selective aromatase inhibitor, which inhibits human placental aromatase with an IC50 of 15 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9896 Supplier Medchemexpress LLC Supplier No. HY1427410MG
Medchemexpress, HY-14274 10mg Anastrozole CAS:120511-73-1 Anastrozole is a potent, highly selective aromatase inhibitor, which inhibits human placental aromatase with an IC50 of 15 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.