Learn More
Apexbio Technology LLC Anastrozole 120511-73-1 200mg
SDP
Catalog No. 5000089788
Encompass_Preferred
Anastrozole (CAS 120511-73-1) is a selective non-steroidal aromatase inhibitor that suppresses estrogen biosynthesis by inhibiting the aromatase enzyme In in vitro assays using human placental aromatase anastrozole demonstrated an IC50 of 14 6 nM (0 0043 g/ml) Oral administration at 0 1 mg/kg completely inhibited ovulation in preclinical models and eliminated exogenous androgen-induced uterotrophic activity in juvenile rodents Additionally it has been shown to inhibit the conversion of 11-deoxycortisol to cortisol in guinea pig dog and bovine adrenal microsomes with IC50 values of 4 09 M 129 M and 11 9 M respectively Anastrozole is widely used in research to study estrogen-mediated pathways and endocrine modulation
Catalog No. 50-000-89788 Supplier Apexbio Technology LLC Supplier No. B1382200
Anastrozole (CAS 120511-73-1) is a selective non-steroidal aromatase inhibitor that suppresses estrogen biosynthesis by inhibiting the aromatase enzyme In in vitro assays using human placental aromatase anastrozole demonstrated an IC50 of 14 6 nM (0 0043 g/ml) Oral administration at 0 1 mg/kg completely inhibited ovulation in preclinical models and eliminated exogenous androgen-induced uterotrophic activity in juvenile rodents Additionally it has been shown to inhibit the conversion of 11-deoxycortisol to cortisol in guinea pig dog and bovine adrenal microsomes with IC50 values of 4 09 M 129 M and 11 9 M respectively Anastrozole is widely used in research to study estrogen-mediated pathways and endocrine modulation