Learn More
Medchemexpress LLC Anastrozole | 120511-73-1 | MFCD00866298 | >98.0% | C17H19N5 | 10MG
SDP
Catalog No. 5000261377
Encompass_Preferred
Anastrozole (Standard) is an analytical reference standard of the non-steroidal aromatase inhibitor anastrozole, provided for research and analytical applications. It is intended for use in method development, qualitative and quantitative assays, and instrument calibration.
  • Potent, selective aromatase inhibitor (IC50 ~15 nM).
  • Intended for analytical use in HPLC, GC, and MS.
  • High purity suitable for analytical assays (>98.0% by HPLC).
  • Available in multiple milligram pack sizes for method development.
  • CAS registry number 120511-73-1 for unambiguous identification.
  • Used for calibration, assay validation, and quantitative analysis.
Catalog No. 50-002-61377 Supplier Medchemexpress LLC Supplier No. HY14274R10MG
Anastrozole (Standard) is an analytical reference standard of the non-steroidal aromatase inhibitor anastrozole, provided for research and analytical applications. It is intended for use in method development, qualitative and quantitative assays, and instrument calibration.
  • Potent, selective aromatase inhibitor (IC50 ~15 nM).
  • Intended for analytical use in HPLC, GC, and MS.
  • High purity suitable for analytical assays (>98.0% by HPLC).
  • Available in multiple milligram pack sizes for method development.
  • CAS registry number 120511-73-1 for unambiguous identification.
  • Used for calibration, assay validation, and quantitative analysis.