Learn More
Medchemexpress LLC Anastrozole-d12 | 120512-32-5 | 99.7% | C17H7D12N5 | 1 MG
SDP
Catalog No. 5000355640
Encompass_Preferred
Anastrozole-d12 is the deuterium labeled Anastrozole, a potent and highly selective aromatase inhibitor. It inhibits human placental aromatase with an IC50 of 15 nM. This compound is intended for research use only and can serve as a tracer or an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuterium substitution may impact the pharmacokinetic and metabolic profiles of drugs. It appears as a white to off-white solid.
  • Potent and highly selective aromatase inhibitor
  • Inhibits human placental aromatase with an IC50 of 15 nM
  • Deuterium labeled Anastrozole
  • Can be used as a tracer for quantitative analysis
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • White to off-white solid appearance
  • Deuterium substitution can affect pharmacokinetic and metabolic profiles
Catalog No. 50-003-55640 Supplier Medchemexpress LLC Supplier No. HY14274S1MG
Anastrozole-d12 is the deuterium labeled Anastrozole, a potent and highly selective aromatase inhibitor. It inhibits human placental aromatase with an IC50 of 15 nM. This compound is intended for research use only and can serve as a tracer or an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuterium substitution may impact the pharmacokinetic and metabolic profiles of drugs. It appears as a white to off-white solid.
  • Potent and highly selective aromatase inhibitor
  • Inhibits human placental aromatase with an IC50 of 15 nM
  • Deuterium labeled Anastrozole
  • Can be used as a tracer for quantitative analysis
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • White to off-white solid appearance
  • Deuterium substitution can affect pharmacokinetic and metabolic profiles