Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Anatabine dicitrate | 2741481-51-4 | MFCD32690113 | 99.0% | 544.46 | C22H28N2O14 | 10 MG
SDP

Catalog No. 5000297004
Encompass_Preferred

Anatabine dicitrate is a tobacco-derived alkaloid research reagent that functions as a potent α4β2 nicotinic acetylcholine receptor agonist and an inhibitor of NF-κB signaling. It can cross the blood-brain barrier and has been used in preclinical studies to reduce amyloid-β production and modulate neuroinflammatory pathways.

  • Potent α4β2 nAChR agonist and NF-κB inhibitor.
  • Demonstrates ability to lower amyloid-β production in models.
  • Can cross the blood-brain barrier.
  • High purity (approximately 99.0%).
  • Solid, white to off-white appearance.
  • Molecular weight 544.46; formula C22H28N2O14.
  • Available in small research pack sizes such as 10 MG.

Catalog No. 50-002-97004 Supplier Medchemexpress LLC Supplier No. HY19918A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Anatabine dicitrate is a tobacco-derived alkaloid research reagent that functions as a potent α4β2 nicotinic acetylcholine receptor agonist and an inhibitor of NF-κB signaling. It can cross the blood-brain barrier and has been used in preclinical studies to reduce amyloid-β production and modulate neuroinflammatory pathways.

  • Potent α4β2 nAChR agonist and NF-κB inhibitor.
  • Demonstrates ability to lower amyloid-β production in models.
  • Can cross the blood-brain barrier.
  • High purity (approximately 99.0%).
  • Solid, white to off-white appearance.
  • Molecular weight 544.46; formula C22H28N2O14.
  • Available in small research pack sizes such as 10 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.