Learn More
Medchemexpress LLC Anatabine dicitrate | 99.03% | 544.46 | 50 MG
SDP
Catalog No. 5000402020
Encompass_Preferred
Anatabine dicitrate is a tobacco alkaloid capable of crossing the blood-brain barrier. It acts as a potent α4β2 nAChR agonist. This compound inhibits NF-κB activation, which leads to a reduction in amyloid-β (Aβ) production by preventing the β-cleavage of amyloid precursor protein (APP). Additionally, Anatabine dicitrate demonstrates anti-inflammatory effects and shows potential for the treatment of neurodegenerative disorders.
  • Can cross the blood-brain barrier.
  • Potent α4β2 nAChR agonist.
  • Inhibits NF-κB activation.
  • Lowers amyloid-β (Aβ) production by preventing the β-cleavage of APP.
  • Possesses anti-inflammatory effects.
  • Has potential for neurodegenerative disorders treatment.
Catalog No. 50-004-02020 Supplier Medchemexpress LLC Supplier No. HY19918A50MG
Anatabine dicitrate is a tobacco alkaloid capable of crossing the blood-brain barrier. It acts as a potent α4β2 nAChR agonist. This compound inhibits NF-κB activation, which leads to a reduction in amyloid-β (Aβ) production by preventing the β-cleavage of amyloid precursor protein (APP). Additionally, Anatabine dicitrate demonstrates anti-inflammatory effects and shows potential for the treatment of neurodegenerative disorders.
  • Can cross the blood-brain barrier.
  • Potent α4β2 nAChR agonist.
  • Inhibits NF-κB activation.
  • Lowers amyloid-β (Aβ) production by preventing the β-cleavage of APP.
  • Possesses anti-inflammatory effects.
  • Has potential for neurodegenerative disorders treatment.