Learn More
TARGETMOL CHEMICALS INC ANATIBANT 2HCL 5MG
SDP
Catalog No. 502580683
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Anatibant 2HCl is a small molecule bradykinin B2 receptor antagonist for the treatment of neurological disorders and may be used in the study of brain injury. purity: 99%
Catalog No. 50-258-0683 Supplier TARGETMOL CHEMICALS INC Supplier No. T26627L5MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Anatibant 2HCl is a small molecule bradykinin B2 receptor antagonist for the treatment of neurological disorders and may be used in the study of brain injury. purity: 99%