Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Anatibant hydrochloride | 209788-45-4 | 99.3% | 1 MG
SDP

Catalog No. 5000233548
Encompass_Preferred

Anatibant hydrochloride is the hydrochloride salt of Anatibant, a selective non-peptide bradykinin B2 receptor antagonist used in preclinical research. It is supplied as an off-white to light yellow solid in small research pack sizes and is accompanied by analytical and safety documentation.

  • Selective bradykinin B2 receptor antagonist.
  • Used for research into brain injury and related pathologies.
  • Crosses the blood-brain barrier.
  • Provided as the dihydrochloride salt.
  • High purity, approximately 99.3%.
  • Molecular weight 784.58 g/mol.
  • CAS number 209788-45-4.
  • Analytical and safety documents available (data sheet, COA, NMR, LCMS, SDS).

Catalog No. 50-002-33548 Supplier Medchemexpress LLC Supplier No. HY121156A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Anatibant hydrochloride is the hydrochloride salt of Anatibant, a selective non-peptide bradykinin B2 receptor antagonist used in preclinical research. It is supplied as an off-white to light yellow solid in small research pack sizes and is accompanied by analytical and safety documentation.

  • Selective bradykinin B2 receptor antagonist.
  • Used for research into brain injury and related pathologies.
  • Crosses the blood-brain barrier.
  • Provided as the dihydrochloride salt.
  • High purity, approximately 99.3%.
  • Molecular weight 784.58 g/mol.
  • CAS number 209788-45-4.
  • Analytical and safety documents available (data sheet, COA, NMR, LCMS, SDS).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.