Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anatomical Chart Series - Diseases, Disorders, and Processes
SCI_ED

Catalog No. S9695DU Shop All Anatomical Chart Company Products
Change view
Click to view available options
Product Type:
Disorders of the Eye Anatomical Chart
Hip/Knee Inflammation Anatomical Chart
Middle Ear Infections Chart
Skin and Common Disorders Chart
Understanding Breast Disease Anatomical Chart
Understanding Ulcers Anatomical Chart
Format:
Laminated
Unmounted
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Format
S9695DU Disorders of the Eye Anatomical Chart Unmounted
S24217PL Skin and Common Disorders Chart Laminated
S9871DU Hip/Knee Inflammation Anatomical Chart Unmounted
S9731PL Understanding Ulcers Anatomical Chart Laminated
S9892PL Middle Ear Infections Chart Laminated
S9757DU Understanding Breast Disease Anatomical Chart Unmounted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. S9695DU Supplier Anatomical Chart Company Supplier No. 9695DU
Add to Cart
Add to Cart

Setting the highest standard of detail found in anatomical charts today!

Extraordinary, highly detailed charts ideal for school use.

Both the 11 x 14 in. desk size and the 20 x 26 in. poster size are available unmounted or laminated; laminated charts come with a special water-soluble marking pen as a teaching aid.

Specifications

Product Type Disorders of the Eye Anatomical Chart
Format Unmounted
For Use With (Application) Extraordinary, highly detailed charts ideal for school use
Subject Science
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.