Learn More
Electron Microscopy Sciences Anatomy Dissecting Kit 2, 1 set
SDP
Catalog No. 50365945
Encompass_Preferred
Kit includes cartilage knife 2" blade, dissecting scalpel handle #4, dissecting scalpel handle #3, scalpel blade #10, 1 piece, scalpel blade #24, 1 piece, dissecting scissor, 5.5", sharp/blunt points, iris dissecting scissors, 4.5, dissecting teasing needle, straight, metal chuc, dissecting teasing needle, angular, metal chuck, dissecting forceps 4.5 with guide pin, fine points, dissecting forceps 5, probe and hook, chrome, ruler 6 plastic, metric/English markings, dissecting chain & hook chrome, dissecting blow pipe, 6, and double folded, vinyl storage case.
Catalog No. 50-365-945 Supplier Electron Microscopy Sciences Supplier No. 7295130
Kit includes cartilage knife 2" blade, dissecting scalpel handle #4, dissecting scalpel handle #3, scalpel blade #10, 1 piece, scalpel blade #24, 1 piece, dissecting scissor, 5.5", sharp/blunt points, iris dissecting scissors, 4.5, dissecting teasing needle, straight, metal chuc, dissecting teasing needle, angular, metal chuck, dissecting forceps 4.5 with guide pin, fine points, dissecting forceps 5, probe and hook, chrome, ruler 6 plastic, metric/English markings, dissecting chain & hook chrome, dissecting blow pipe, 6, and double folded, vinyl storage case.