Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Learning Resources Anatomy Models Set
SCI_ED

Catalog No. S95867
Change view
Click to view available options
Product Type:
Anatomical Model, Brain
Anatomical Model, Heart
Anatomical Model, Human Body
Model, Anatomy Set
Model, Human Skeleton
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S95867 Anatomical Model, Brain
S95868 Anatomical Model, Human Body
S95866 Anatomical Model, Heart
S95870 Model, Anatomy Set
S95869 Model, Human Skeleton
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options

CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.

Catalog No. S95867 Supplier Learning Resources, Inc Supplier No. LER3335
Add to Cart
Add to Cart

CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.

Helps students gain a deeper understanding of how organs and systems interact.

Ideal for human body centers focusing on major organs or the circulatory, nervous or skeletal systems. Each realistically detailed plastic model includes a display stand and guide with facts and step-by-step, photo-illustrated assembly instructions.

Brain (S95867)

  • Features cerebellum, frontal lobe, parietal lobe, temporal lobe, occipital lobe, corpus callosum, brain stem, hippocampus, ventricles, insula, corpus striatum, internal capsule and lentiform nucleus
  • 31-piece model measures 3.75 in.H (9.75cm) when assembled

Heart (S95866)

  • Features superior vena cava, inferior vena cava, right atrium, tricuspid valve, right ventricle, pulmonary valve, pulmonary artery, left atrium, mitral valve, left ventricle, aortic valve and aorta
  • Opens when assembled so students can explore from the inside
  • 29-piece model measures 5 in.H (12.7cm) when assembled

Human Body (S95868)

  • Features brain, skull, heart, rib cage, lungs, liver, stomach, pancreas, kidneys, small intestine, large intestine and spine
  • 31-piece torso model measures 4.5 in.H (11.4cm) when assembled

Skeleton (S95869)

  • Features skull, rib cage, humerus, spinal column, radius, ulna, hand, pelvis, femur, tibia, fibula, foot and partial circulatory system
  • 41-piece model measures 9.2 in.H (23.4cm) when assembled

Order Info

Grades 3+.

Specifications

Dimensions (L x W x H) 28.8 x 14.6 x 3.8 in. (73.15 x 37 x 9.65 cm)
For Use With (Application) For deeper understanding of how the parts of the brain work together
Height (English) 3.8 in.
Height (Metric) 9.65 cm
Includes Cerebellum, frontal, parietal, temporal and occipital lobes, corpus callosum, brain stem, hippocampus, ventricles, insula, corpus striatum, internal capsule and lentiform nucleus, fact guide and photo-illustrated assembly instructions
Length (English) 28.8 in.
Length (Metric) 73.15 cm
Material Plastic
Mounting Display Stand
Width (English) 14.6 in.
Width (Metric) 37 cm
Product Type Anatomical Model, Brain
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.