Learn More
Sheldon Manufacturing Inc ANATOX REFILL 3LB GRANULES
SDP
Catalog No. NC0106683
Encompass_Preferred
Anaerobic activated carbon, 2lbs. For use with Bactron and Bactrox chambers to mitigate VOC and VSC build-up
Catalog No. NC0106683 Supplier Sheldon Manufacturing Inc Supplier No. 1060500
Anaerobic activated carbon, 2lbs. For use with Bactron and Bactrox chambers to mitigate VOC and VSC build-up