Learn More
Enzo Life Sciences (+/-) Anatoxin (1mg). CAS: 64285-06-9
SDP
Supplier:  Enzo Life Sciences BMLC1180001
Encompass_Preferred
(±)2-Acetyl-Anatoxin is an alkaloidal toxin produced by the filamentous freshwater cyanophyte Anabaena flos-aquae.  This compound is the synthetic version. Purity: ≥98% (TLC). Solubility: Soluble in water (14mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0344