Learn More
Apexbio Technology LLC (±)-Anatoxin A fumarate 1219922-30-1 1mg
SDP
Catalog No. 5000091905
Encompass_Preferred
( )-Anatoxin A fumarate (CAS 1219922-30-1) is a naturally occurring alkaloid isolated from freshwater cyanobacteria It functions as an agonist at nicotinic acetylcholine receptors exhibiting high binding affinity for rat brain 4 2 (K i 1 25 nM) and lower affinity for 7 (K i 1 84 M) subtypes In functional assays ( )-Anatoxin A fumarate induces dopamine release in rat striatal synaptosomes with an EC 50 of 134 nM This compound is utilized in neuroscience research to investigate cholinergic signaling and dopaminergic mechanisms in the central nervous system
Catalog No. 50-000-91905 Supplier Apexbio Technology LLC Supplier No. B64531
( )-Anatoxin A fumarate (CAS 1219922-30-1) is a naturally occurring alkaloid isolated from freshwater cyanobacteria It functions as an agonist at nicotinic acetylcholine receptors exhibiting high binding affinity for rat brain 4 2 (K i 1 25 nM) and lower affinity for 7 (K i 1 84 M) subtypes In functional assays ( )-Anatoxin A fumarate induces dopamine release in rat striatal synaptosomes with an EC 50 of 134 nM This compound is utilized in neuroscience research to investigate cholinergic signaling and dopaminergic mechanisms in the central nervous system