Learn More
Grainger Prime-Line Zip-It Wall Anchors, Nylon, #8, 50/Pk.
Catalog No. 502791417
Encompass_Preferred
These wall anchors are constructed of heavy-duty nylon to provide a secure anchor point in hollow walls. The self-drilling design allows for efficient installation without the need for pre-drilling.
  • Constructed of durable nylon
  • Designed for heavy-duty applications
  • Provides secure anchoring in hollow wall surfaces
  • Features a self-drilling design for easy installation
  • Compatible with #8 screw sizes
  • Supplied in a 50-pack
Catalog No. 50-279-1417 Supplier Grainger Supplier No. 169X74
These wall anchors are constructed of heavy-duty nylon to provide a secure anchor point in hollow walls. The self-drilling design allows for efficient installation without the need for pre-drilling.
  • Constructed of durable nylon
  • Designed for heavy-duty applications
  • Provides secure anchoring in hollow wall surfaces
  • Features a self-drilling design for easy installation
  • Compatible with #8 screw sizes
  • Supplied in a 50-pack